Assay Details
Gene Symbol: jpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY394952.1 | - | 2 - 3 | 327 | 63 | |
BC045436.1 | - | 2 - 3 | 306 | 63 | ||
BC066731.1 | - | 2 - 3 | 232 | 63 | ||
BC155180.1 | - | 2 - 3 | 251 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
