Assay Details
Gene Symbol: cs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199598.1 | NP_955892.1 | 5 - 6 | 508 | 75 | |
GenBank mRNA | BC045362.1 | - | 5 - 6 | 508 | 75 | |
BC165743.1 | - | 5 - 6 | 438 | 75 | ||
BC166040.1 | - | 5 - 6 | 504 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
