Assay Details
Gene Symbol: eef1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199949.2 | NP_956243.1 | 2 - 3 | 488 | 127 | |
GenBank mRNA | AY398339.1 | - | 2 - 3 | 488 | 127 | |
BC046042.1 | - | 2 - 3 | 271 | 127 | ||
BC071464.1 | - | 2 - 3 | 273 | 127 | ||
BC150470.1 | - | 2 - 3 | 272 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
