Assay Details
Gene Symbol: napsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200031.1 | NP_956325.1 | 2 - 3 | 390 | 69 | |
GenBank mRNA | BC056836.1 | - | 2 - 3 | 390 | 69 | |
BC154324.1 | - | 2 - 3 | 322 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
