Assay Details
Gene Symbol: vps29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200037.2 | NP_956331.1 | 2 - 3 | 295 | 65 | |
XM_005161202.4 | XP_005161259.1 | 3 - 4 | 298 | 65 | ||
GenBank mRNA | BC045981.1 | - | 2 - 3 | 297 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
