Assay Details
Gene Symbol: znf131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200505.2 | NP_956799.2 | 3 - 4 | 713 | 62 | |
XM_021478213.1 | XP_021333888.1 | 3 - 4 | 396 | 62 | ||
XM_021478215.1 | XP_021333890.1 | 3 - 4 | 396 | 62 | ||
GenBank mRNA | BC044506.1 | - | 3 - 4 | 336 | 62 | |
BC063944.1 | - | 3 - 4 | 713 | 62 | ||
BC164150.1 | - | 2 - 3 | 256 | 62 |
Gene Symbol: znf346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC055532.1 | - | 3 - 4 | 514 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
