Assay Details
Gene Symbol: ppa1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200733.1 | NP_957027.1 | 5 - 6 | 410 | 73 | |
GenBank mRNA | BC059512.1 | - | 5 - 6 | 410 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
