Assay Details
Gene Symbol: trappc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200764.1 | NP_957058.1 | 3 - 4 | 480 | 100 | |
GenBank mRNA | BC059594.1 | - | 3 - 4 | 480 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
