Assay Details
Gene Symbol: prdx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200805.1 | NP_957099.1 | 4 - 5 | 578 | 76 | |
GenBank mRNA | BC059671.1 | - | 4 - 5 | 578 | 76 | |
BC153616.1 | - | 4 - 5 | 581 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
