Assay Details
Gene Symbol: ccl19a.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082906.2 | NP_001076375.2 | 3 - 4 | 372 | 79 | |
XM_005165054.1 | XP_005165111.1 | 5 - 6 | 711 | 79 | ||
GenBank mRNA | AB331770.1 | - | 3 - 4 | 372 | 79 | |
BC122386.1 | - | 5 - 6 | 714 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
