Assay Details
Gene Symbol: dnase2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114738.1 | NP_001108210.1 | 5 - 6 | 597 | 103 | |
XM_021466569.1 | XP_021322244.1 | 5 - 6 | 647 | 103 | ||
GenBank mRNA | BC159268.1 | - | 5 - 6 | 607 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
