Assay Details
Gene Symbol: cxl34b.11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115062.1 | NP_001108534.1 | 4 - 5 | 319 | 69 | |
GenBank mRNA | AB331756.1 | - | 4 - 5 | 319 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
