Assay Details
Gene Symbol: mrpl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122621.1 | NP_001116093.1 | 6 - 7 | 431 | 104 | |
GenBank mRNA | BC159118.1 | - | 6 - 7 | 431 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
