Assay Details
Gene Symbol: nck2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200997.1 | NP_957291.1 | 3 - 4 | 1122 | 90 | |
XM_009304471.3 | XP_009302746.1 | 4 - 5 | 1386 | 90 | ||
XM_021478633.1 | XP_021334308.1 | 3 - 4 | 2864 | 90 | ||
GenBank mRNA | BC046872.1 | - | 3 - 4 | 1122 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
