Assay Details
Gene Symbol: amd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161601.1 | NP_001155073.1 | 7 - 8 | 945 | 102 | |
GenBank mRNA | BC045404.1 | - | 7 - 8 | 924 | 102 | |
BC163959.1 | - | 7 - 8 | 742 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
