Assay Details
Gene Symbol: nedd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201321.2 | NP_958478.2 | 3 - 4 | 297 | 80 | |
GenBank mRNA | BC055645.1 | - | 3 - 4 | 294 | 80 | |
EH443336.1 | - | 3 - 4 | 323 | 80 | ||
EH507246.1 | - | 3 - 4 | 300 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
