Assay Details
Gene Symbol: ap1s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205558.1 | NP_991121.1 | 3 - 4 | 336 | 149 | |
GenBank mRNA | BC044496.1 | - | 3 - 4 | 362 | 149 | |
BC056547.1 | - | 3 - 4 | 336 | 149 | ||
BC065471.1 | - | 3 - 4 | 336 | 149 | ||
BC155185.1 | - | 3 - 4 | 347 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
