Assay Details
Gene Symbol: gria1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205598.1 | NP_991161.1 | 13 - 14 | 2442 | 103 | |
GenBank mRNA | AF525741.1 | - | 13 - 14 | 2442 | 103 | |
BC162505.1 | - | 13 - 14 | 2378 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
