Assay Details
Gene Symbol: wbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205634.1 | NP_991197.1 | 1 - 2 | 185 | 91 | |
XM_017356050.2 | XP_017211539.1 | 1 - 2 | 285 | 91 | ||
GenBank mRNA | BC065355.1 | - | 1 - 2 | 185 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
