Assay Details
Gene Symbol: brd3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212702.1 | NP_997867.1 | 10 - 11 | 2057 | 100 | |
XM_009301940.3 | XP_009300215.1 | 11 - 12 | 2178 | 100 | ||
GenBank mRNA | BC065949.1 | - | 10 - 11 | 2057 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
