Assay Details
Gene Symbol: brd3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212702.1 | NP_997867.1 | 9 - 10 | 1767 | 85 | |
XM_005165593.4 | XP_005165650.2 | 10 - 11 | 1902 | 85 | ||
XM_005165594.4 | XP_005165651.2 | 10 - 11 | 1885 | 85 | ||
XM_005165595.4 | XP_005165652.2 | 9 - 10 | 1819 | 85 | ||
XM_009301936.3 | XP_009300211.1 | 10 - 11 | 1888 | 85 | ||
XM_009301938.3 | XP_009300213.1 | 9 - 10 | 1819 | 85 | ||
XM_009301940.3 | XP_009300215.1 | 10 - 11 | 1888 | 85 | ||
GenBank mRNA | BC065949.1 | - | 9 - 10 | 1767 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
