Assay Details
Gene Symbol: slc16a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212708.1 | NP_997873.1 | 3 - 4 | 419 | 122 | |
XM_005169469.4 | XP_005169526.1 | 3 - 4 | 614 | 122 | ||
GenBank mRNA | AB610608.1 | - | 2 - 3 | 393 | 122 | |
BC045489.1 | - | 3 - 4 | 419 | 122 | ||
BC154231.1 | - | 3 - 4 | 419 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
