Assay Details
Gene Symbol: esrra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212955.1 | NP_998120.1 | 3 - 4 | 622 | 109 | |
XM_005161344.4 | XP_005161401.1 | 4 - 5 | 874 | 109 | ||
XM_009295473.3 | XP_009293748.1 | 5 - 6 | 1178 | 109 | ||
GenBank mRNA | AY556395.1 | - | 3 - 4 | 622 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
