Assay Details
Gene Symbol: rsl24d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212993.1 | NP_998158.1 | 2 - 3 | 262 | 87 | |
GenBank mRNA | BC051780.1 | - | 2 - 3 | 262 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
