Assay Details
Gene Symbol: ppib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213019.1 | NP_998184.1 | 2 - 3 | 411 | 81 | |
GenBank mRNA | BC059560.1 | - | 2 - 3 | 278 | 81 | |
BC071458.1 | - | 2 - 3 | 411 | 81 | ||
BC154213.1 | - | 2 - 3 | 302 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
