Assay Details
Gene Symbol: dda1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213127.1 | NP_998292.1 | 3 - 4 | 361 | 107 | |
XM_005155700.4 | XP_005155757.1 | 4 - 5 | 327 | 107 | ||
GenBank mRNA | BC054628.1 | - | 3 - 4 | 361 | 107 | |
BC155199.1 | - | 3 - 4 | 344 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
