Assay Details
Gene Symbol: rps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213377.1 | NP_998542.1 | 4 - 5 | 380 | 126 | |
GenBank mRNA | AY648802.1 | - | 4 - 5 | 378 | 126 | |
BC046054.1 | - | 4 - 5 | 380 | 126 | ||
BC159194.1 | - | 4 - 5 | 380 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
