Assay Details
Gene Symbol: nudt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213418.1 | NP_998583.1 | 4 - 5 | 432 | 92 | |
GenBank mRNA | BC049486.1 | - | 4 - 5 | 432 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
