Assay Details
Gene Symbol: calm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199996.2 | NP_956290.1 | 5 - 6 | 467 | 145 | |
GenBank mRNA | BC059427.1 | - | 5 - 6 | 467 | 145 | |
BC068339.1 | - | 5 - 6 | 467 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
