Assay Details
Gene Symbol: UBE2Q2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489739.6 | XP_001489789.2 | 1149 | 114 | ||
XM_005602881.3 | XP_005602938.2 | 1036 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
