Assay Details
Gene Symbol: FRS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501157.5 | XP_001501207.1 | 442 | 85 | ||
XM_023624916.1 | XP_023480684.1 | 1627 | 85 | |||
XM_023624917.1 | XP_023480685.1 | 392 | 85 | |||
XM_023624918.1 | XP_023480686.1 | 532 | 85 | |||
XM_023624920.1 | XP_023480688.1 | 631 | 85 | |||
XM_023624921.1 | XP_023480689.1 | 280 | 85 | |||
XM_023624922.1 | XP_023480690.1 | 457 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
