Assay Details
Gene Symbol: DOP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650712.1 | XP_023506480.1 | 493 | 69 | ||
XM_023650713.1 | XP_023506481.1 | 541 | 69 | |||
XM_023650715.1 | XP_023506483.1 | 386 | 69 | |||
XM_023650716.1 | XP_023506484.1 | 493 | 69 | |||
XM_023650717.1 | XP_023506485.1 | 493 | 69 | |||
XM_023650718.1 | XP_023506486.1 | 493 | 69 | |||
XM_023650719.1 | XP_023506487.1 | 493 | 69 | |||
XM_023650720.1 | XP_023506488.1 | 314 | 69 | |||
XM_023650721.1 | XP_023506489.1 | 493 | 69 | |||
XM_023650722.1 | XP_023506490.1 | 314 | 69 | |||
XM_023650723.1 | XP_023506491.1 | 493 | 69 | |||
XM_023650724.1 | XP_023506492.1 | 314 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
