Assay Details
Gene Symbol: EXTL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495867.5 | XP_001495917.1 | 3026 | 110 | ||
XM_023636221.1 | XP_023491989.1 | 3382 | 110 | |||
XM_023636222.1 | XP_023491990.1 | 3257 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
