Assay Details
Gene Symbol: VIRMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489304.5 | XP_001489354.1 | 3831 | 108 | ||
XM_014728200.2 | XP_014583686.1 | 3831 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
