Assay Details
Gene Symbol: CAB39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494866.6 | XP_001494916.2 | 847 | 153 | ||
XM_023642533.1 | XP_023498301.1 | 996 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() |
