Assay Details
Gene Symbol: RXRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014734342.2 | XP_014589828.1 | 907 | 87 | ||
XM_014734344.2 | XP_014589830.1 | 666 | 87 | |||
XM_023624702.1 | XP_023480470.1 | 1274 | 87 | |||
XM_023624703.1 | XP_023480471.1 | 1274 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
