Assay Details
Gene Symbol: GZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014735010.2 | XP_014590496.1 | 1729 | 88 | ||
XM_023625994.1 | XP_023481762.1 | 1559 | 88 | |||
XM_023625995.1 | XP_023481763.1 | 1826 | 88 | |||
XM_023625996.1 | XP_023481764.1 | 1621 | 88 | |||
XM_023625997.1 | XP_023481765.1 | 1562 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
