Assay Details
Gene Symbol: KMT5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654594.1 | XP_023510362.1 | 997 | 88 | ||
XM_023654595.1 | XP_023510363.1 | 879 | 88 | |||
XM_023654596.1 | XP_023510364.1 | 889 | 88 | |||
XM_023654597.1 | XP_023510365.1 | 928 | 88 | |||
XM_023654598.1 | XP_023510366.1 | 512 | 88 | |||
XM_023654599.1 | XP_023510367.1 | 432 | 88 | |||
XM_023654600.1 | XP_023510368.1 | 997 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
