Assay Details
Gene Symbol: SUPT7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005600113.3 | XP_005600170.1 | 1436 | 93 | ||
XM_005600114.3 | XP_005600171.1 | 1457 | 93 | |||
XM_023619325.1 | XP_023475093.1 | 1436 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
