Assay Details
Gene Symbol: OGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499535.5 | XP_001499585.1 | 788 | 73 | ||
XM_023640514.1 | XP_023496282.1 | 437 | 73 | |||
XM_023640519.1 | XP_023496287.1 | 2057 | 73 | |||
GenBank mRNA | JL631787.1 | - | 3 - 4 | 183 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
