Assay Details
Gene Symbol: PDGFC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500645.6 | XP_001500695.1 | 777 | 67 | ||
XM_023636394.1 | XP_023492162.1 | 791 | 67 | |||
XM_023636395.1 | XP_023492163.1 | 381 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
