Assay Details
Gene Symbol: BABAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502017.5 | XP_001502067.1 | 1241 | 70 | ||
XM_023619317.1 | XP_023475085.1 | 1132 | 70 | |||
GenBank mRNA | JL616309.1 | - | 11 - 12 | 1170 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
