Assay Details
Gene Symbol: PRKAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163958.1 | NP_001157430.1 | 4 - 5 | 1058 | 144 | |
XM_014742136.2 | XP_014597622.1 | 744 | 144 | |||
GenBank mRNA | DQ318137.1 | - | 4 - 5 | 1058 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
