Assay Details
Gene Symbol: TBCCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309330.1 | NP_001296259.1 | 4 - 5 | 1085 | 93 | |
XM_014732634.2 | XP_014588120.1 | 1210 | 93 | |||
XM_014732635.2 | XP_014588121.1 | 1245 | 93 | |||
XM_023623139.1 | XP_023478907.1 | 1517 | 93 | |||
XM_023623140.1 | XP_023478908.1 | 1517 | 93 | |||
XM_023623141.1 | XP_023478909.1 | 1517 | 93 | |||
XM_023623142.1 | XP_023478910.1 | 1563 | 93 | |||
XM_023623143.1 | XP_023478911.1 | 1214 | 93 | |||
XM_023623144.1 | XP_023478912.1 | 1222 | 93 | |||
XM_023623145.1 | XP_023478913.1 | 2980 | 93 | |||
GenBank mRNA | JL640600.1 | - | 1 - 2 | 43 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
