Assay Details
Gene Symbol: DDX3X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023634084.1 | XP_023489852.1 | 1165 | 54 | ||
XM_023634085.1 | XP_023489853.1 | 1167 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
