Assay Details
Gene Symbol: RAB27B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001487851.6 | XP_001487901.1 | 647 | 110 | ||
XM_014727868.2 | XP_014583354.1 | 677 | 110 | |||
XM_023647829.1 | XP_023503597.1 | 722 | 110 | |||
XM_023647830.1 | XP_023503598.1 | 476 | 110 | |||
XM_023647831.1 | XP_023503599.1 | 859 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
