Assay Details
Gene Symbol: GRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283084.1 | NP_001270013.1 | 1 - 2 | 186 | 74 | |
XM_001495766.6 | XP_001495816.1 | 455 | 74 | |||
GenBank mRNA | HQ890091.1 | - | 1 - 2 | 186 | 74 | |
HQ890180.1 | - | 1 - 2 | 186 | 74 | ||
JL615874.2 | - | 1 - 2 | 190 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
