Assay Details
Gene Symbol: POLD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100414.2 | NP_001093884.2 | 4 - 5 | 335 | 64 | |
GenBank mRNA | AB331246.1 | - | 4 - 5 | 343 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
