Assay Details
Gene Symbol: HNRNPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023637848.1 | XP_023493616.1 | 997 | 105 | ||
XM_023637849.1 | XP_023493617.1 | 852 | 105 | |||
XM_023637850.1 | XP_023493618.1 | 997 | 105 | |||
XM_023637851.1 | XP_023493619.1 | 852 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
