Assay Details
Gene Symbol: CNOT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301266.2 | NP_001288195.1 | 2 - 3 | 432 | 75 | |
XM_005610612.3 | XP_005610669.1 | 357 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
